Learn More
Corning™ 96-Well Clear-Bottom Nonbinding Surface (NBS™) Microplates Available on GSA/VA Contract for Federal Government customers only.
Enhanced signal-to-noise ratio
Manufacturer:  Corning™ 3604
 View more versions of this product
Catalog No. 07-200-732
Add to cart
Description
Description
  • Ideal for homogenous assays such as Scintillation Proximity Assays (SPA) and fluorescent assays
  • Modified polymer surface is nonionic hydrophilic, minimizing bio-interaction within wells to reduce nonspecific binding and reagent costs
  • Significantly reduces protein and nucleic acid binding to polymers, maintains enzyme activity and inhibits adhesion of a number of cell lines
  • Stable to aqueous solutions containing EtOH
  • Use at temperature range of 4° to 37°C (39° to 99°F)
  • Flat-bottom wells
  • Compatible with a wide range of plate readers, liquid handlers and robotic systems
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
White
Plate readers, liquid handlers and robotic systems
NBS™
4° to 37°C
75 to 200μL
Non-sterile
For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
96 well
39° to 99°F
360μL
Clear