Learn More
Corning™ 96-Well Half Area Black Flat Bottom Polystyrene Microplates
Designed to reduce well-to-well crosstalk
Manufacturer:  Corning™ 3721
 View more versions of this product
Catalog No. 07-200-815
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
  • Choice of NBS™ or TC-treated surface coating
Specifications
Specifications
Flat
Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
Tissue-Culture Treated
25 to 125μL
96-Well Assay Microplate
Clear
Polystyrene
190μL
With Lid
Sterile