Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion
Analyze endothelial cell invasion using fluorescence detection in a simplified and reproducible manner
Manufacturer:  Corning™ 354142
 View more versions of this product
Catalog No. 08-774-379
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
  • Allows invasion of Human Microvascular Endothelial Cells (HMVEC-1) - a human microvascular endothelial cell line
  • Excludes invasion of 3T3 cells - a non-invasive fibroblast cell line
Specifications
Specifications
24
Membrane
5/Pk.
3μm
Endothelial Cell Invasion