Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation
Evaluate and quantitate endothelial cell invasion using real-time fluorescence and non-destructive sample preparation
Manufacturer:  Corning™ 354149
 View more versions of this product
Catalog No. 08-774-376
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Includes: A Falcon™ 24-Multiwell Insert Plate and a non-tissue culture-treated 24-well receiver plate with lid
Description
Description
·    In vitro assay eliminates the need for manual cell scraping and counting
·    Insert plate contains a fluorescence blocking microporous (3.0 µm pore size) polyethylene terephthalate (PET) membrane (BD FluoroBlok™) coated with BD Matrigel™ Matrix
·    BD Matrigel Matrix blocks the passage of non-invasive cells while allowing the passage of invasive activated endothelial cells
·    Robust system allows IC50 measurement
Specifications
Specifications
24
3μm
Microplate
1