Learn More
Corning BioCoat Angiogenesis System for Endothelial Cell Tube Formation
Evaluate and quantitate endothelial cell invasion using real-time fluorescence and non-destructive sample preparation
$455.00 - $1964.00
Specifications
Pore Size | 3 μm |
---|---|
Type | Microplate |
No. of Wells | 96 |
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel, BioCoat, FluoroBlok and PureCoat) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Includes: A Falcon 24-Multiwell Insert Plate and a non-tissue culture-treated 24-well receiver plate with lid
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
08-774-376
|
Corning
354149 |
1 |
Each for $455.00
|
|
08-774-377
|
Corning
354150 |
5 |
Case of 5 for $1,964.00
|
|
Description
- Highly reproducible assay system
- Ideal for screening anti-angiogenic agents
- Rapid image acquisition and data quantification
Specifications
3 μm | |
Microplate | |
96 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.