Learn More
Corning™ BioCoat™ Collagen I Multiwell Plates
Treated with BioCoat™ for enhanced cell attachment and growth of a variety of primary cells and ransformed cell lines in serum-free or reduced serum cultures. Corning™ BioCoat™ Collagen I Multiwell Plates closely mimmics in vivo environments for cell culture applications. Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
Manufacturer:  Corning™ 354397
 View more versions of this product
Catalog No. 08-775-32
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
  • Source: Rat tail
  • TC-treated
  • Clear, round well
  • Flat bottom
  • Nonsterile
For research use only.
Specifications
Specifications
ANSI, SBS
Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
Polystyrene
Rat Tail
ECM/Attachment Factors
With Lid
384-Well Collagen I Multiwell Microplate, Low Volume
Black
384 well
5/Cs.
4° to 30°C
Clear
Round
Non-sterile
Safety and Handling
Safety and Handling
ShelfLife : 6 Months