Corning™ BioCoat™ Collagen I Multiwell Plates

Treated with BioCoat™ for enhanced cell attachment and growth of a variety of primary cells and ransformed cell lines in serum-free or reduced serum cultures. Corning™ BioCoat™ Collagen I Multiwell Plates closely mimmics in vivo environments for cell culture applications. Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

Manufacturer: Corning™ 356702

 View more versions of this product

Catalog No. 10-130-3

  Check Availability
Add to Cart

Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


    Specifications

    Lid Yes
    Format 96-well
    Material Polystyrene
    Quantity 5/Cs.
    Shelf Life 6 Months
    Source Rat Tail
    Sterile No
    Storage Requirements 4° to 30°C
    Surface Coating ECM/Attachment Factors,ECM/Attachment Factors
    Type 96-Well Collagen I Multiwell Microplate
    Well Color Clear,Clear
    Color Clear
    For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
    Certifications/Compliance ANSI, SBS
    Well Design Round
    Closure Yes,Yes
    Well Shape Round
    Bottom Plate Material Polystyrene
    Applications Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture

    • Source: Rat tail
    • TC-treated
    • Clear, round well
    • Flat bottom
    • Nonsterile

    For research use only.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit