Learn More
Corning BioCoat Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning BioCoat Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.
$91.30 - $1099.00
Specifications
Closure Material | Phenolic |
---|---|
Barcode | Yes |
Closure Type | Vented |
Surface Coating | Collagen I |
For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel, BioCoat, FluoroBlok and PureCoat) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Catalog Number | Mfr. No. | Capacity (Metric) | Culture Area | No. per Case | Neck Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Culture Area | No. per Case | Neck Style | Price | Quantity & Availability | ||
08-774-329
|
Corning
354486 |
600 mL | 150 cm2 | 5/Cs. | Canted |
Case of 5 for $173.50
|
|
08-774-328
|
Corning
354485 |
250 mL | 75 cm2 | 5/Cs. | Canted |
Case of 5 for $91.30
|
|
08-774-327
![]() |
Corning
354484 |
70 mL | 25 cm2 | 10/Cs. | Canted |
Case of 10 for $147.60
|
|
08-774-330
|
Corning
354487 |
750 mL | 175 cm2 | 5/Cs. | Straight |
Case of 5 for $185.50
|
|
08-774-336
![]() |
Corning
356484 |
70 mL | 25 cm2 | 50/Cs. | Canted |
Case of 50 for $431.50
|
|
08-774-338
![]() |
Corning
356486 |
600 mL | 150 cm2 | 40/Cs. | Canted |
Case of 40 for $927.00
|
|
08-774-339
![]() |
Corning
356487 |
750 mL | 175 cm2 | 40/Cs. | Straight |
Case of 40 for $1,099.00
|
|
08-774-337
![]() |
Corning
356485 |
250 mL | 75 cm2 | 50/Cs. | Canted |
Case of 50 for $641.00
|
|
Description
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
Applications include:
- Promotion of cell attachment and spreading
- Rapid expansion of cell populations
- Serum-free or reduced serum culture
- Cell adhesion assays
- Improving survival of primary cells in culture
Source
Quality Control
- Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
- Tested and found negative for bacteria and fungi
- Collagen I purity >90% by SDS-PAGE
Storage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Phenolic | |
Vented | |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
Sterile |
Yes | |
Collagen I | |
Rectangular |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.