Learn More
Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.
Manufacturer:  Corning™ 354487
 View more versions of this product
Catalog No. 08-774-330
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
  • Vented caps allow gas exchange and minimize risk of contamination
  • Ideal for adhesion and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
Specifications
Specifications
Yes
Phenolic
175cm2
For adhesion, growth and differentiation of neuronal and transfected cell lines
5/Cs.
Sterile
25.3 oz.
Vented
750mL
Canted
Rectangular
Collagen I