Learn More
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Manufacturer:  Corning™ 354459
 View more versions of this product
Catalog No. 08-774-31
Add to cart
Description
Description
Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo.
  • Source: Mouse
  • TC-treated
  • Nonsterile
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
Specifications
Collagen IV
35mm
Round (U)
Polystyrene