Learn More
Corning™ BioCoat™ Control Insert
Use alongside BD BioCoat™ inserts when studying effects of the ECM component present on the BD BioCoat cell culture insert
Manufacturer:  Corning™ 354569
 View more versions of this product
Catalog No. 08-774-157
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
Each lot tested and found negative for bacteria and fungi
    • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology Supplied with preapplied extracellular matrices, facilitating cell adaption to in vitro cultivation
    • Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces
    • Low pore-density, track-etched polyethylene teraphthalate (PET) membranes for convenient viewing of living cells with a phase-contrast microscope
    • Have low-binding properties that minimize small molecule loss
    These products are for research use only and are not intended for diagnostic purposes.
    Order Info
    Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
Specifications
Specifications
12
1μm
2
24
Well Plate
2° to 8°C