Learn More
Corning BioCoat Fibronectin HTS Multiwell Angiogenesis
Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays
$3260.00
Specifications
Pore Size | 3 μm |
---|---|
Type | Multiwell Insert Plate |
For Use With (Application) | High Throughput Screening |
Surface Coating | Fibronectin |
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel, BioCoat, FluoroBlok and PureCoat) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Includes: A Falcon 24- and 96-Multiwell insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid
Catalog Number | Mfr. No. | Culture Area | No. of Wells | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Culture Area | No. of Wells | Quantity | Price | Quantity & Availability | |||
08-774-399
|
Corning
354148 |
0.08 cm2 | 96 | 5 |
Case of 5 for $3,260.00
|
|
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
3 μm | |
Multiwell Insert Plate | |
High Throughput Screening | |
Fibronectin |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.