Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis

Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays

Manufacturer:  Corning™ 354147

 View more versions of this product

Catalog No. 08-774-398


Add to cart

Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Includes: A Falcon 24- and 96-Multiwell™ insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid

Description

Description

  • Combines the fluorescence blocking capabilities of the BD Falcon™ FluoroBlok™ membrane with the benefits of an extracellular matrix protein
  • Provides a predictive in vitro model for screening pro- and anti-angiogenic agents
  • Eliminates tedious manual cell scraping and counting
  • Fibronectin concentration and coating procedure have been optimized so the pores of the membrane are not obstructed, allowing endothelial cells to attach to the membrane and freely migrate toward an angiogenic stimulus in the receiver plate
  • Cells may be labeled with a fluorescent dye either pre- or post-migration and quantified using a fluorescence microplate reader
  • Light-tight membrane blocks the transmission of light within the 490 to 700nm range, shielding fluorescently labeled cells in the top of the insert from detection by a bottom-reading microplate reader and allowing for real-time detection without dismantling the insert
  • Tested for the ability to support migration of HUVEC (human umbilical vein endothelial cells) in response to VEGF (vascular endothelial cell growth factor)
  • Automation compatible
  • Stable for at least three months when stored at -20°C

Specifications

Specifications

96
3μm
High Throughput Screening
Fibronectin
0.08 cm2
Multiwell Insert Plate
1
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit