Learn More
Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis
Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays
Supplier: Corning™ 354148
Includes: A Falcon 24- and 96-Multiwell™ insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
0.08 cm2 | |
3 μm | |
High Throughput Screening | |
5/Pk. | |
Fibronectin |
PET | |
Multiwell Insert Plate | |
5 | |
5/Cs. | |
96 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.