Learn More
Corning™ BioCoat™ Fibronectin Multiwell Plate
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
Manufacturer:  Corning™ 354501
 View more versions of this product
Catalog No. 08-774-57
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology Supplied with preapplied extracellular matrices, facilitating cell adaption to in vitro cultivation
  • Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo
  • Low pore-density, track-etched polyethylene teraphthalate (PET) membranes offer convenient viewing of living cells with a phase-contrast microscope
  • Have low-binding properties that minimize small molecule loss
The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody. These products are for research use only and are not intended for diagnostic purposes.
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
Specifications
Specifications
Clear
Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays
Polystyrene
2° to 8°C
Clear
Round
Non-sterile
3.8cm2
12 well
Human Fibronectin
ECM/Attachment Factors
With Lid
12-Well Fibronectin Multiwell Microplate
Safety and Handling
Safety and Handling
ShelfLife : 3 Months