Learn More
Corning™ BioCoat™ Fibronectin Rectangular Canted Neck Culture Flask
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
Manufacturer:  Corning™ 354646
 View more versions of this product
Catalog No. 08-774-217
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
  • Matrix components are applied as an even, optically clear coating
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody
Specifications
Specifications
20.2 oz.
Plug-seal
600mL
Canted
Sterile
Phenolic
150cm2
For adhesion, growth and differentiation of neuronal and transfected cell lines
Rectangular
Fibronectin