Learn More
Corning BioCoat Fibronectin TC-treated Culture Dish
Matrix components are applied as an even, optically clear coating
$195.30 - $198.10
Specifications
Bottom Shape | Round |
---|---|
Material | Polystyrene |
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel, BioCoat, FluoroBlok and PureCoat) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
08-774-63
|
Corning
354451 |
100 mm |
Case of 10 for $198.10
|
|
08-774-62
|
Corning
354403 |
60 mm |
Case of 20 for $195.30
|
|
08-774-61
|
Corning
354457 |
35 mm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
08-774-64
|
Corning
354552 |
150 mm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody.
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Round | |
Polystyrene |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.