Learn More
Corning™ BioCoat™ Matrigel™ Matrix Thin Layer Culture Dishes
Tested for the ability to promote neurite outgrowth from chick dorsal root ganglia (in the absence of NGF)
Manufacturer:  Corning™ 354602
 View more versions of this product
Catalog No. 08-774-180
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
  • Soluble basement membrane extracted from Engelbreth-Holm-Swarm mouse tumor
  • When reconstituted at room temperature, forms a gel
  • Especially suited for culture of polarized cells, such as epithelial cells, and promotes differentiation of hepatocytes, mammary epithelial cells and smooth muscle cells
  • Also promotes capillary tubule formation of endothelial cells and neurite regeneration, and can be used for angiogenesis assays
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
35mm
Round (U)
Matrigel Membrane