Learn More
Corning™ BioCoat™ Matrigel Matrix Multiwell Plates
Cell culture microplate
$576.00 - $591.50
Specifications
Material Polystyrene
Closure Included Yes
For Use With (Application) Elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
Color Clear
Sterility Non-sterile
View More Specs
Includes: Lid
Products 2
Catalog Number Mfr. No. Well Count Price Quantity & Availability  
Catalog Number Mfr. No. Well Count Price Quantity & Availability  
08-774-107
View Documents Promotion Details
Corning™
354432
6
Case of 2 for $576.00
Add to cart
08-774-109
View Documents Promotion Details
Corning™
354433
24
Case of 2 for $591.50
Add to cart
Description
Description
For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.
Specifications
Specifications
Polystyrene
Elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
Non-sterile
ECM/Attachment Factors
Yes
Clear
-20°C
Rectangular
Safety and Handling
Safety and Handling
ShelfLife : 3 Months