Learn More
Corning™ BioCoat™ Matrigel Matrix Multiwell Plates
Cell culture microplate
Manufacturer:  Corning™ 354432
 View more versions of this product
Catalog No. 08-774-107
Add to cart
Includes: Lid
Description
Description
Clear, flat bottom
TC-treated
Nonsterile
Specifications
Specifications
Clear
Elicitation of tissue-specific cellular morphology and protein production in epithelial cells, differentiation of endothelial, muscle and neuronal cells, development of three-dimensional matrix model systems
Polystyrene
EHS Mouse Tumor
ECM/Attachment Factors
With Lid
6-Well Matrix Multiwell Microplate
9.6cm2
6 well
6
-20°C
Clear
Round
Non-sterile
Safety and Handling
Safety and Handling
ShelfLife : 3 Months