Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid; Type: Optical Glass Base

Manufacturer:  Corning™ 4587

Catalog No. 15-100-165


Add to cart

Description

Description

High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.

  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 μm is well suited for imaging microscopy.
  • Bottom flatness <50 μm to ensure planarity for imaging devices
  • Low background fluorescence and minimal cross-talk provide the highest possible optical quality for cell-based assays.
Specifications

Specifications

Half-Area
1/Pk., 10/Cs.
Flat
10/Cs.
Non-sterile
384
With Lid
Optical Glass Base
Black, Clear Bottom
1/Pk.
Poly-D Lysine-Treated
16 x 24
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit