Learn More
Corning™ BioCoat™ Poly-L-Lysine Culture Dishes
Provide excellent substrate for cell attachment and differentiation of primary neurons, glial, neuroblastoma and glioma cells
Manufacturer:  Corning™ 356518
 View more versions of this product
Catalog No. 08-772-95
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
Suitable for promoting adhesion of transfected cell lines and other fastidious cell types. Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology.
  • Minimize assay failure due to cell detachment
  • Matrix components are applied as an even, optically clear coating
  • Precoated with Poly-L-Lysine to eliminate time-consuming preparation
  • Lot-to-lot consistency provides reproducible experiments
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
Poly-L-Lysine
35mm
Round (U)