Learn More
Corning™ BioCoat™ Tumor Invasion Multiwell Plate Available on GSA/VA Contract for Federal Government customers only.
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
Manufacturer:  Corning™ 354166
 View more versions of this product
Catalog No. 08-774-381
Add to cart
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
Description
·    Ideal for studies of the metastatic potential of tumor cells and the inhibition of invasion by antineoplastic drugs
·    Saves time and labor when screening for prospective antimetastatic compounds
·    Eliminates tedious manual cell scraping and counting
·    Includes a 24- or 96-Multiwell™ insert plate containing an 8.0µm pore size FluoroBlok™ polyethylene terephthalate membrane coated with BD Matrigel™ Matrix; the matrix provides a true barrier to noninvasive cells while presenting an appropriate protein structure for invading cells to penetrate and pass through the FluoroBlok membrane
·    Light-tight membrane ensures that only those cells that appear on the underside of the insert are detected by a fluorescence microplate reader, providing a signal that can be directly correlated to cell number
·    Each lot is tested for the ability to allow invasion of HT-1080 cells (an invasive human fibrosarcoma cell line) and to exclude invasion of 3T3 cells (a noninvasive fibroblast cell line)
·    Automation compatible
·    Store at -20°C
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
24
PET
Multiwell Plate
0.3 cm2
8μm
5