Learn More
Corning™ BioCoat™ Tumor Invasion Multiwell Plate
GSA_VA
Provides an environment that allows for the assessment of the invasive potential of cells in vitro
$3930.00
Specifications
Material PET
Pore Size 8 μm
Type Multiwell Plate
View More Specs
Products 4
Catalog Number Mfr. No. Culture Area No. of Wells Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Culture Area No. of Wells Quantity Price Quantity & Availability  
08-772-62
View Documents
Corning™
354168
0.08 cm2 96 5
Case of 5 for $3,930.00
Add to cart
08-772-61
GSA_VA
View Documents
Corning™
354167
0.08 cm2 96 1 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
08-774-380
View Documents
Corning™
354165
0.3 cm2 24 1 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
08-774-381
GSA_VA
View Documents
Corning™
354166
0.3 cm2 24 5 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running cell migration and invasion studies.
Specifications
Specifications
PET
8 μm
Multiwell Plate