Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners Available on GSA/VA Contract for Federal Government customers only.
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
Manufacturer: Corning™ 999813
 View more versions of this product
Catalog No. 02-883-8C
  Check Availability
Add to Cart
    Specifications
    Color Black
    Liner Material PTFE, Rubber
    Product Type Screw Cap
    Height (Metric) 13.5mm
    Thread Size 13-415
    Tip Material PTFE, Rubber
    Autoclavable Yes
    Material Phenolic
    • Glued-in rubber lining firmly bonded to an interface of PTFE
    • Autoclavable and reusable
  • Description & Specifications
Related Products
Product Content Feedback