Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Cell Culture Untreated Flasks

Printed with lot numbers for ease in traceability

Manufacturer:  Corning™ 431464

 View more versions of this product

Catalog No. 08-757-501

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

2.02 oz.
HDPE
75cm2
Yes
Canted
Sterile
60mL
Orange
Vented
For optimal cell attachment
Lot numbers for ease in traceability
Rectangular
60mL
PS (Polystyrene)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit