Learn More
Corning™ Cell Scrapers Available on GSA/VA Contract for Federal Government customers only.
Angled swivel blade minimizes cell damage and ensures even contact with growth area. Corning™ Cell Scrapers are ideal for harvesting cells.
Manufacturer:  Corning™ 3011
 View more versions of this product
Catalog No. 07-200-366
Add to cart
Description
Description
Angled swivel blade minimizes cell damage and ensures even contact with growth area. Corning™ Cell Scrapers are ideal for harvesting cells.
  • Long handle permits easy access to cell culture flasks and roller bottles 
  • Designed for use in 25, 75 and 225cmflasks
  • Sterile; certified nonpyrogenic
  • Individually wrapped
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Sterile
3cm
39cm
Blade with handle
1.18 in.
15 in.
Cell Scraper
Documents
Accessories