Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ CellBIND™ Surface Culture Dishes
Manufactured from optically-clear virgin polystyrene for excellent visibility. Corning™ CellBIND™ Suface Culture Dishes are treated for optimal cell attachment to help cells adapt to reduced serum or serum-free media.
Supplier: Corning™ 3294
Description
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment
- Ultra-Low Attachment surface dishes feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation
- 10 dishes/bag are available for 100 mm dishes (Cat. No. 430293)
- 245 mm square dishes offer 500 cm2 growth surface
- Stacking beads aid in handling
- Vents provide consistent gas exchange
- Manufactured from optically clear virgin polystyrene
- Sterile
- Nonpyrogenic
Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.
Cat. no. 431110, Square dishes with interior bottom dimensions of 224 x 224 mm.
Specifications
Clear | |
Polystyrene | |
2.7 mL (Well) | |
8 cm2 | |
Polystyrene | |
Sterile | |
Cell Culture Petri Dish | |
210/Cs. | |
10 mm |
Yes | |
Round | |
Clear | |
Single-use | |
Round | |
CellBIND | |
Lid | |
10/Pk. | |
35 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction