Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates permit top or bottom analysis using detection microplate readers.
Manufacturer: Corning™ 3683
 View more versions of this product
Catalog No. 05-538-90
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Lid Yes,Yes
    Culture Area 0.06cm2
    Format 384-well,384-well
    Material Polystyrene,Polystyrene
    No. of Wells 384
    Sterile Yes
    Volume (Metric) 112μL,112μL
    Volume (Metric) Working 20 to 80μL
    Well Color Clear,Clear
    Closure Yes
    Color Black/Clear Bottom
    Wells 384
    • Medical-grade polystyrene
    • Well volume: 112µL
    • Working volume: 20µL to 80µL
    • Treated surface options (medium bind, high bind, TC-treated) are not coated, require no special handling and are stable at room temperature
    • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
    • Plate dimensions meet proposed industry standards
  • Description & Specifications
Related Products
Product Content Feedback