Corning™ CellBIND™ Surface Cell Culture Flasks

Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.

Manufacturer: Corning™ 3290CONV

 View more versions of this product

Catalog No. 05-539-104

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Neck Style Canted
    Capacity (English) 2.02 oz.
    Capacity (Metric) 60mL
    Culture Area 75cm2
    Surface Coating Corning™ Cellbind™
    Barcode No
    Capacity 2.02 oz., 60mL
    Closure Color Orange
    Closure Material HDPE
    For Use With (Application) For optimal cell attachment
    Graduated Yes
    Shape Rectangular
    Sterile Yes
    Volume (Metric) Working 15 to 22.5mL
    Closure Type Vented
    Material Polystyrene
    Includes Lot numbers for ease in traceability

    • Manufactured from optically clear, virgin polystyrene
    • Treated surface helps cells adapt to reduced serum or serum-free media; stable at room temperature
    • Ideal for short- and long-term cell culture
    • Available with vented caps containing a 0.2µm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination; or phenolic-style caps for use in open systems requiring gas exchange
    • 3290 flask features redesigned neck for easier media pouring, lower height (1.4 in.) for reduced stacking height, and vented cap with Costar™-style concentric circle support grid geometry
    • Choice of canted or angled neck
      • Canted neck flasks allow easier pouring and improved access to the flask for pipetting or scraping
      • Angled neck improves pipetting access and reduces medium sloshing into the neck
    • Printed lot numbers provide easy traceability
    • Sterilized by gamma irradiation
    • Certified nonpyrogenic
    • 100% integrity tested

    • Flasks 431328 and 431346 are prelabeled with bar code for use with SelecT™ Robotic System

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit