Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CellSTACK™

Products meet a minimum Sterility Assurance Level (SAL) of 10-6

$2630.00 - $5250.00

Specifications

Description Nonpyrogenic
Length (Metric) 335 mm
Width (Metric) 206 mm
View More Specs

Products 2
Catalog Number Mfr. No. Culture Area Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Culture Area Height (Metric) Price Quantity & Availability  
10-320-195
View Documents
Corning™
11874
1272 cm² 31 mm
Case for $5,250.00
Add to cart
 
10-320-196
View Documents
Corning™
11875
6360 cm² 203 mm
Case for $2,630.00
Add to cart
 
Description

Description

  • Product has been sterilized and dosimetrically released 
  • Products meet a minimum Sterility Assurance Level (SAL) of 10-6
  • Non-Pyrogenic
  • Bacterial Endotoxins test methodologies, routine monitoring, and alternative to batch testing
  • Tested for the attribute of cell attachment and growth utilizing an attachment-dependent mammalian cell line
Specifications

Specifications

Nonpyrogenic
335 mm
206 mm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit