Corning™ General Assay Microplate Lids

Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.

Manufacturer: Corning™ 3095

 View more versions of this product

Catalog No. 07-200-375

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    For Use With All 96-well
    No. per Case 100
    No. per Pack 100.0
    Sterile No
    Material Acetate
    Format Sealing Tape

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit