Learn More
Novus Biologicals™ Corning Costar Spin-X Centrifuge Tube Filters
0.22µm pore cellulose acetate membrane-containing filter unit within a 500µL capacity polypropylene microcentrifuge tube; ideal for removing bacteria, particles, or cells from liquids, HPLC sample preparation and DNA removal from agarose or acrylamide gels
Manufacturer:  Novus Biologicals™ SPINX8160016NOV
 View more versions of this product
Catalog No. SPINX816001
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Corning Costar™ Spin-X Centrifuge Tube Filters
Store at room temperature.
16 Each