Corning™ Costar™ Tip Loading System Starter Kit
Graduations and superb optical clarity offer easy volume identification
Manufacturer: Corning™ 4781
 View more versions of this product
Catalog No. 07-200-781
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Volume (Metric) 1 to 200μL
    Color Yellow
    • Statistical Process Control (SPC) used in the molding operation gives Costar pipet tips consistent straightness, length, and sealing ring location
    • Tips properly attach to and eject from pipetter every time
    • Minimal dimensional tolerances deliver specific volumes with reliable accuracy
    • Choose from filtered IsoTip and Universal Fit Tips
    • Available bulk-packed and compatible with a wide array of pipetters
    • Universal Fit Tips also available racked
    • For easy loading of tip racks, see the unique Tip Loading System
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback