Corning™ Cryogenics Workstation Available on GSA/VA Contract for Federal Government customers only.
Rack grips Corning externally and internally threaded self-standing vials, so caps can be removed with one hand
Manufacturer: Corning™ 430526
 View more versions of this product
Catalog No. 03-374-37B
  Check Availability
Add to Cart
    Specifications
    Product Type Cryogenic Vial Rack
    Holds 30 cryogenic vial
    Material Polycarbonate
    Color White
  • Description & Specifications
Related Products
Product Content Feedback