Learn More
Corning™ Untreated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
Manufacturer:  Corning™ 430588
 View more versions of this product
Catalog No. 08-772-30
Add to cart
Description
Description
  • Manufactured from optically-clear virgin polystyrene
  • Not treated for cell attachement
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Sterile
10mm
9cm2
35mm
Optically Clear Virgin Polystyrene (PS)