Corning™ DeckWorks Pipet Tips Available on GSA/VA Contract for Federal Government customers only.

Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking

Manufacturer: Corning™ 4119

 View more versions of this product

Catalog No. 10-320-708

  Check Availability
Add to Cart

    Specifications

    Product Type Standard Tip
    Sterility Nonsterile
    Tip Style Hinged Rack, Universal Fit
    Shape Round
    Packaging Hinged Rack
    For Use With (Application) Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
    Volume (Metric) 100 to 1000μL
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit