Corning™ DeckWorks™ Reload Systems with Standard Tips Available on GSA/VA Contract for Federal Government customers only.
Packaged in transparent trays for easy tip identification. Corning™ Deckworks™ Reload Systems with Standard Tips provide convenient and economical reloading of durable hinged racks.
Manufacturer: Corning™ 4126
 View more versions of this product
Catalog No. 10-320-715
  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Standard Tip
    Material Polypropylene
    Quantity 960 tips/Pk.; 4 Pack/Cs.
    Sterility Nonsterile
    Graduated Yes
    Autoclavable Yes
    Tip Style Standard
    Shape Round
    Packaging Deck
    Volume (Metric) 1 to 200μL
    Color Natural
    • Available in 10µL, 200µL, 300µL and 1,000µL sizes
    • Requires no transfer devices, saves space and minimize packaging waste
    • Pipet tips are made from virgin medical-grade resins
    • Reload decks and rack bases are manufactured from recycled polypropylene
    • Nonsterile decks and racks are steam autoclavable
  • Description & Specifications
Related Products
Product Content Feedback