Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Disposable 115mL Sterile Vacuum Filter Unit

Features low center of gravity and wide base for stability

Manufacturer:  Corning™ 430945

 View more versions of this product

Catalog No. 09-761-95B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: 60mm diameter membrane

Description

Description

  • No-mess pour spout saves time and makes sample transfer easier
  • Eliminates need to detach tubing to empty filtered media, decreasing chance of sample contamination
  • Porosity is clearly labeled on each unit
  • Certified nonpyrogenic
  • Sterile
  • Cellulose acetate (CA) membrane for low protein binding
  • Available in 0.22 (for sterilizing) or 0.45µm (for clarifying) pore sizes

Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit