Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Disposable Spinner Flask with Top Cap and Angled Sidearms Available on GSA/VA Contract for Federal Government customers only.

Ready-to-use system comes with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly

Manufacturer:  Corning™ 3153

 View more versions of this product

Catalog No. 07-201-153


Add to cart

Description

Description

  • Made of ISO 10993 polystyrene
  • Comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads
  • Paddle size and height are optimized for each vessel size
  • Magnet provides a smooth, even rotation at required speeds on any laboratory slow speed stirrer
  • Heat buildup in the vessel is reduced by means of a specially designed flange that raises the vessel off the stir-plate surface
  • Flared base provides extra stability and support

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

0.132 gal.
Polypropylene
3.8cm
4.5cm
PS (Polystyrene)
8 in.
100mm Top cap and 2 Angled side arms
2
GL45
Orange
1.5 in.
87.3mm
500mL
Yes
20.3cm
45°
Sterile
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit