Learn More
Corning™ DNA-BIND™ 96-Well Plates
Convenient method for covalently immobilizing ssDNA by either the 5« or 3« end
Supplier: Corning™ 2505
Description
- Ready to use without surface preparation, prehybridization, or addition of expensive crosslinking reagents or highly purified oligos
- Ideal for hybridization and amplification
- Surface provides binding with high specificity and extremely low background
- Flat-bottom wells
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
Non-sterile | |
Ideal for hybridization and amplification | |
1/Pk. | |
Clear |
Without Lid | |
Clear | |
96 | |
Round | |
50/Cs. | |
50/Cs. |