Learn More
Corning™ Epoxide-Coated Microarray Slides
Ideal for covalent attachment of unmodified or amino-modified short oligonucleotides (~30-mer) and long oligonucleotides (>50-mer)
Manufacturer:  Corning™ 40044
 View more versions of this product
Catalog No. 07-202-524
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
No
Epoxide
25 Case
75.5mm
1.1mm
Microarray Slide
Significant advantage in oligonucleotide retention and post-hybridization signal strength, for covalent attachment of unmodified or amino-modified short oligonucleotides (∼30 mer) and long oligonucleotides (>50 mer)
25.3mm
0.04 in.
No