Corning™ FiltrEX Filter Plates Available on GSA/VA Contract for Federal Government customers only.
Individual filter discs eliminate liquid cross-contamination
Manufacturer: Corning™ 3511
 View more versions of this product
Catalog No. 07-200-764
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Ideal for automated liquid handling, centrifugation and vacuum filtration, wide skirt easily accommodates bar code labels
    Format 96-well
    Material Polystyrene
    Membrane Hydrophilic PVDF
    No. of Wells 96
    Pore Size 0.66mm
    Sterile No
    Type Filtering Microplate
    Certifications/Compliance ANSI, SBS1
    Volume (Metric) Well 350μL,350μL
    Color White
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback