Learn More
Corning™ Hybridization Chamber
Designed to hold 25 x 75mm microarray slides at a constant humidity during hybridization incubations
Manufacturer:  Corning™ 2551
 View more versions of this product
Catalog No. 07-200-271
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Polycarbonate and nonsterile
  • O-ring and retaining clips assure the reusable chamber remains watertight in waterbaths and airtight in hybridization ovens
  • Wells in the base hold 10µL of water to maintain optimal interior humidity
  • Increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
  • Used with raised-edge coverslips that are thicker or taller than standard thin coverglass
Specifications
Specifications
Hybridization Chamber
Wells in the base hold 10μL of water to maintain optimal interior humidity, increased interior depth allows for single slide hybridizations and for user to place two arrays face-to-face and hybridize using a single labeled target
5 Case
Polycarbonate