Learn More
Corning™ HYPERStack™ Accessories: Stack Manipulator Available on GSA/VA Contract for Federal Government customers only.
Accessories for use with Corning HYPERStack products
Manufacturer:  Corning™ 11000
Catalog No. 07-201-851
Add to cart
This item is not returnable. View return policy
Description
Description
  • Made of stainless steel for durability
  • Accommodates both HYPERStack-36 and CellSTACK-10 vessels
  • Ergonomic design
  • Easy dials for protocol development
  • Handle two units simultaneously or one unit at a time
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Stainless Steel
1/Cs.
Stack manipulator
Hyperstack™ 110and Hyperstack™ 36 Vessels
1/Pk.