Learn More
Corning™ HYPERStack™ Culture Vessels Available on GSA/VA Contract for Federal Government customers only.
Closed system that allows fluid manipulations to occur through preassembled tubing sets
Manufacturer:  Corning™ 10037
 View more versions of this product
Catalog No. 07-201-872
Add to cart
Description
Description
  • 2.5 times more cells per volumetric footprint
  • Closed system, no open fluid manipulations
  • Scalable, multiple size offerings
  • Ergonomic design for easier manipulation
  • Fixed media volume – 0.2 mL/cm2 fills vessel for optimal handling
  • Innovative Assembly
  • No adhesives, low particulate
  • Lower disposal Ccost
  • Less volumetric waste per growth area
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Sterile
18000cm2
36 layer vessel; non-treated
2 Case
Gamma sterilized to SAL 10-6 standard