Corning™ Internally Threaded Cryogenic Vials with Color Caps Available on GSA/VA Contract for Federal Government customers only.

Used for ultra-low temperature storage of valuable cell cultures, biological solutions and aqueous solutions

Manufacturer: Corning™ 431421

 View more versions of this product

Catalog No. 13-700-505

  Check Availability
Add to Cart

    Specifications

    Product Type Cryogenic Vial
    Closure Material Polypropylene
    DNase Free Yes
    Thread Style Internal
    Sterile Yes
    Color Natural
    For Use With (Application) For ultra low temperature storage of valuable cell cultures, biological solutions and aqueous solutions
    Graduated Yes
    Includes Silicone washer, inserts
    Label Type Marking Spot
    Packaging 500/Case
    Quantity 50/Pk.
    Shape Round
    RNase Free Yes
    Temperature Range (Metric) −196°C
    Temperature Range (English) −320.8°F
    Color Closure White
    Material Polypropylene
    Capacity 2mL

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit