Learn More
Corning™ Lambda Plus 8 and 12-Channel Pipettor Available on GSA/VA Contract for Federal Government customers only.
Designed for universal fit with all common brands of pipet tips
Manufacturer:  Corning™ 4083
 View more versions of this product
Catalog No. 07-764-709
Add to cart
Description
Description
  • Features smooth plunger movement and low pipetting forces
  • Volume can be set by turning either thumb wheel or the plunger button
  • Accidental change prevented by locking mechanism
  • Key included for in-house calibration
  • Available in 8 and 12 channels
  • Available in 1 to 300µL volume
  • Adjustable tip ejector
  • Fully autoclavable and UV resistant
  • Factory calibrated to strict ISO 8655 standards
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Accessory stand and rack available separately (Cat. Nos. 07-201-790 and -789).
Warranty and Services
3-Year
Specifications
Specifications
±1.6 to ±1%
CE
200/300μL
3 Years
50 to 300μL
Yes
Non-sterile
Yes
8