Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Low Profile Cell Culture Flask

Unique design reduces stacking height

Manufacturer:  Corning™ 3816

 View more versions of this product

Catalog No. 07-200-878

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • 100cm2 available growth area
  • Rectangular with canted neck, corner cap for easy pouring and improved access to flask for pipetting or scraping
  • Manufactured from optically clear virgin polystyrene
  • Available with patented Corning CellBIND™ surface treatment for enhanced cell attachment or tissue culture treatment
  • Vented cap contains 0.2µm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination
  • Graduated
  • 100% integrity tested
  • Sterilized by gamma radiation and certified nonpyrogenic

Specifications

Specifications

7.1 oz.
HDPE
100cm2
PE (Polyethylene)
Yes
Rectangular
Tissue culture treated
Orange
Vented
210mL
For optimal cell attachment and growth
Canted
Sterile
210mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit