Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Matrigel™ Membrane Matrix HC Available on GSA/VA Contract for Federal Government customers only.

High concentration (HC) formulation is ideal for applications in which a high protein concentration augments the growth of tumors; allows a Matrigel basement membrane matrix plug to maintain its integrity after subcutaneous injection into mice

Manufacturer:  Corning™ 354263

 View more versions of this product

Catalog No. 08-774-392


Add to cart

Alert:

Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer


Description

Description

Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.

  • Tested for the ability to promote neurite outgrowth of chick dorsal root ganglia cells
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Source: Engelbreth-Holm-Swarm (EHS) mouse tumor
  • Base formulation: Dulbecco's Modified Eagle's Medium with 50µg/mL gentamycin, with or without Phenol Red
  • Solubilized basement membrane preparations polymerize at room temperature

  • Phenol Red-free is available for assays that require color detection, including fluorescence assays

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Sterile
Basement membrane matrix
Engelbreth-Holm-Swarm Mouse Tumor
Growth Factor Reduced (GFR); LDEV-Free
10mL
-20°C
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit